accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1152782-19-8

NFFFOHNOHO

CAS RN 1152782-19-8

IUPAC Name: 2-({4-[2-(trifluoromethyl)phenyl]piperidine-1-carbonyl}amino)benzoic acid
Synonyms: 2-{4-[2-(trifluoromethyl)phenyl]piperidine-1-carbonylamino}benzoic acid
Molecular Weight (g/mol): 392.38
Molecular Formula: C20H19F3N2O3
InChi Key: MEAQCLPMSVEOQF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1NC(=O)N1CCC(CC1)C1=CC=CC=C1C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products