CAS RN 116-06-3
CAS RN 116-06-3
IUPAC Name:
(E)-[2-methyl-2-(methylsulfanyl)propylidene]amino N-methylcarbamate
Synonyms:
temik rcra waste number P070 temik G10 temik G aldicarb (substance) temik 10 G union carbide UC-21149 sulfone aldoxycarb union carbide 21149 ccris 17
Molecular Weight (g/mol):
190.26
Molecular Formula:
C7H14N2O2S
InChi Key:
QGLZXHRNAYXIBU-WEVVVXLNSA-N
SMILES:
CNC(=O)O\N=C\C(C)(C)SC
1
–
6
of
6
results
2
SPEX Certiprep Aldicarb, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
6
Honeywell Fluka Aldicarb, Fluka™
Analytical standard, reference material