accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1160058-86-5

ONNFFFSOOCl

CAS RN 1160058-86-5

IUPAC Name: 4-{[4-(trifluoromethyl)pyrimidin-2-yl]oxy}benzene-1-sulfonyl chloride
Synonyms: 4-{[4-(trifluoromethyl)pyrimidin-2-yl]oxy}benzenesulfonyl chloride
Molecular Weight (g/mol): 338.69
Molecular Formula: C11H6ClF3N2O3S
InChi Key: SMDWXDCIMUGFNP-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NC(OC2=CC=C(C=C2)S(Cl)(=O)=O)=NC=C1

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products