accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1163-19-5

BrBrBrOBrBrBrBrBrBrBr

CAS RN 1163-19-5

IUPAC Name: 1,2,3,4,5-pentabromo-6-(2,3,4,5,6-pentabromophenoxy)benzene
Synonyms: decabromodiphenyl ether
Molecular Weight (g/mol): 959.17
Molecular Formula: C12Br10O
InChi Key: WHHGLZMJPXIBIX-UHFFFAOYSA-N
SMILES: BrC1=C(Br)C(Br)=C(OC2=C(Br)C(Br)=C(Br)C(Br)=C2Br)C(Br)=C1Br
Concentration 
  • (2)
Solvent or Matrix 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products