accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1174064-62-0

NNBrSOOCl

CAS RN 1174064-62-0

IUPAC Name: 4-(4-bromo-1H-pyrazol-1-yl)benzene-1-sulfonyl chloride
Synonyms: 4-(4-bromopyrazol-1-yl)benzenesulfonyl chloride
Molecular Weight (g/mol): 321.57
Molecular Formula: C9H6BrClN2O2S
InChi Key: LRYXAJMNOSSJDD-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(C=C1)N1C=C(Br)C=N1

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products