accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1200-22-2

HOO(R)SS

CAS RN 1200-22-2

IUPAC Name: 5-[(3R)-1,2-dithiolan-3-yl]pentanoic acid
Synonyms: lipoic acid pleomix α pleomix-α DL-α-lipoic acid pyruvate oxidation factor tioctidasi espa-lipon duralipon lipoic acid (substance) trommsdorgg brand of thioctic acid
Molecular Weight (g/mol): 206.32
Molecular Formula: C8H14O2S2
InChi Key: AGBQKNBQESQNJD-SSDOTTSWSA-N
SMILES: OC(=O)CCCC[C@@H]1CCSS1
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products