accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1210-83-9

H3CONHNHOH

CAS RN 1210-83-9

IUPAC Name: N-[2-(5-hydroxy-1H-indol-3-yl)ethyl]acetamide
Synonyms: N-acetylserotonin ASE
Molecular Weight (g/mol): 218.26
Molecular Formula: C12H14N2O2
InChi Key: MVAWJSIDNICKHF-UHFFFAOYSA-N
SMILES: CC(=O)NCCC1=CNC2=CC=C(O)C=C12
Physical Form 
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products