CAS RN 1219728-20-7
CAS RN 1219728-20-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Tocris Bioscience™ SR 7826

Potent and selective LIMK inhibitor; antitumor
2
3