accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 122-75-8

CH3OHOCH3OHOHNNH

CAS RN 122-75-8

IUPAC Name: bis(acetic acid); benzyl[2-(benzylamino)ethyl]amine
Synonyms: bis(acetic acid); benzathine
Molecular Weight (g/mol): 360.45
Molecular Formula: C20H28N2O4
InChi Key: MTRNNCLQPVCDLF-UHFFFAOYSA-N
SMILES: CC(O)=O.CC(O)=O.C(CNCC1=CC=CC=C1)NCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products