CAS RN 135729-62-3
CAS RN 135729-62-3
IUPAC Name:
hydrogen (5R)-3-[(3S)-1-azabicyclo[2.2.2]octan-3-yl]-3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),9,11-trien-2-one chloride
Synonyms:
hydrogen palonosetron chloride
Molecular Weight (g/mol):
332.87
Molecular Formula:
C19H25ClN2O
InChi Key:
OLDRWYVIKMSFFB-KPVRICSOSA-N
SMILES:
[H+].[Cl-].O=C1N(C[C@@H]2CCCC3=CC=CC1=C23)[C@@H]1CN2CCC1CC2
1
–
2
of
2
results
2
Tocris™ Palonosetron hydrochloride
Highly potent and high affinity 5-HT3 antagonist