accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 13709-38-1

FFFLa

CAS RN 13709-38-1

IUPAC Name: lanthanum(3+) trifluoride
Synonyms: lanthanum(3+) trifluoride
Molecular Weight (g/mol): 195.9
Molecular Formula: F3La
InChi Key: BYMUNNMMXKDFEZ-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[La+3]
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)
Grade 
  • (2)
  • (2)

Narrow Results

Narrow Results

  • (10)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products