accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 141-57-1

CH3SiClClCl

CAS RN 141-57-1

IUPAC Name: trichloro(propyl)silane
Synonyms: propyltrichlorosilane
Molecular Weight (g/mol): 177.52
Molecular Formula: C3H7Cl3Si
InChi Key: DOEHJNBEOVLHGL-UHFFFAOYSA-N
SMILES: CCC[Si](Cl)(Cl)Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products