accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 15243-33-1

RuRuRuCCCCCCCCCOOOOOOOOOOCCOCO

CAS RN 15243-33-1

IUPAC Name: dodecakis(methanidylidyneoxidanium) triruthenium
Synonyms: triruthenium dodecacarbonyl
Molecular Weight (g/mol): 639.33
Molecular Formula: C12O12Ru3
InChi Key: NQZFAUXPNWSLBI-UHFFFAOYSA-N
SMILES: [Ru].[Ru].[Ru].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products