CAS RN 1538604-68-0
CAS RN 1538604-68-0
IUPAC Name:
N-(2-{[6-(2,6-dichloro-3,5-dimethoxyphenyl)quinazolin-2-yl]amino}-3-methylphenyl)prop-2-enamide
Synonyms:
N-(2-{[6-(2,6-dichloro-3,5-dimethoxyphenyl)quinazolin-2-yl]amino}-3-methylphenyl)prop-2-enamide
Molecular Weight (g/mol):
509.39
Molecular Formula:
C26H22Cl2N4O3
InChi Key:
TXEBNKKOLVBTFK-UHFFFAOYSA-N
SMILES:
COC1=CC(OC)=C(Cl)C(=C1Cl)C1=CC2=CN=C(NC3=C(C)C=CC=C3NC(=O)C=C)N=C2C=C1
1
–
2
of
2
results
2
Medchem Express BLU9931, MedChemExpress
MedChemExpress BLU9931 is a potent, highly selective, and irreversible fibroblast growth factor receptor 4 (FGFR4) inhibitor with an IC50 of 3 nM and a Kd of 6 nM. BLU9931 has significant antitumor activity.
