accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1726-12-1

H3C

CAS RN 1726-12-1

IUPAC Name: (1-phenylpentyl)benzene
Synonyms: (1-phenylpentyl)benzene
Molecular Weight (g/mol): 224.35
Molecular Formula: C17H20
InChi Key: GSGNVNZDHCKRGI-UHFFFAOYSA-N
SMILES: CCCCC(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products