CAS RN 27470-51-5
CAS RN 27470-51-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Suxibuzone, Mikromolâ„¢
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Medchem Express Suxibuzone, MedChemExpress
MedChemExpress Suxibuzone is a drug used for joint and muscular pain, is a prodrug of the non steroidal anti inflammatory drug Phenylbutazone.
