accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 307538-42-7

H2CNHNNBr

CAS RN 307538-42-7

IUPAC Name: 6-bromo-N-(prop-2-en-1-yl)quinazolin-4-amine
Synonyms: 6-bromo-N-(prop-2-en-1-yl)quinazolin-4-amine
Molecular Weight (g/mol): 264.13
Molecular Formula: C11H10BrN3
InChi Key: BCPOLXUSCUFDGE-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(NCC=C)N=CN=C2C=C1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (6)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products