CAS RN 320-67-2
CAS RN 320-67-2
Thermo Scientific Chemicals 5-Azacytidine, 99%
CAS: 320-67-2 Molecular Formula: C8H12N4O5 Molecular Weight (g/mol): 244.21 MDL Number: MFCD00006539 InChI Key: NMUSYJAQQFHJEW-KVTDHHQDSA-N Synonym: 5-azacytidine,azacitidine,ladakamycin,azacytidine,vidaza,mylosar,azacitidina,azacitidinum,5-azacitidine,azacitidinum inn-latin PubChem CID: 9444 ChEBI: CHEBI:2038 IUPAC Name: 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,3,5-triazin-2-one SMILES: NC1=NC(=O)N(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O
Thermo Scientific Alfa Aesar 5-Azacytidine, 98%, Thermo Scientific™
CAS: 320-67-2 Molecular Formula: C8H12N4O5 Molecular Weight (g/mol): 244.207 MDL Number: MFCD00006539 InChI Key: NMUSYJAQQFHJEW-KVTDHHQDSA-N Synonym: 5-azacytidine,azacitidine,ladakamycin,azacytidine,vidaza,mylosar,azacitidina,azacitidinum,5-azacitidine,azacitidinum inn-latin PubChem CID: 9444 ChEBI: CHEBI:2038 IUPAC Name: 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,3,5-triazin-2-one SMILES: C1=NC(=NC(=O)N1C2C(C(C(O2)CO)O)O)N
MedChemExpress 5-Azacytidine
MedChemExpress 5-Azacytidine (Azacitidine; 5-AzaC; Ladakamycin) is a nucleoside analogue of cytidine that specifically inhibits DNA methylation. 5-Azacytidine is incorporated into DNA to covalently trap DNA methyltransferases and contributes to reverse epigenetic changes. 5-Azacytidine induces cell autophagy.

MedChemExpress 5-Azacytidine
MedChemExpress 5-Azacytidine (Azacitidine; 5-AzaC; Ladakamycin) is a nucleoside analogue of cytidine that specifically inhibits DNA methylation. 5-Azacytidine is incorporated into DNA to covalently trap DNA methyltransferases and contributes to reverse epigenetic changes. 5-Azacytidine induces cell autophagy.

MedChemExpress 5-Azacytidine
MedChemExpress 5-Azacytidine (Azacitidine; 5-AzaC; Ladakamycin) is a nucleoside analogue of cytidine that specifically inhibits DNA methylation. 5-Azacytidine is incorporated into DNA to covalently trap DNA methyltransferases and contributes to reverse epigenetic changes. 5-Azacytidine induces cell autophagy.

MP Biomedicals 5-Azacytidine, MP Biomedicals™
CAS: 320-67-2 Molecular Formula: C8H12N4O5 Molecular Weight (g/mol): 244.21 InChI Key: NMUSYJAQQFHJEW-KVTDHHQDSA-N Synonym: 5-azacytidine,azacitidine,ladakamycin,azacytidine,vidaza,mylosar,azacitidina,azacitidinum,5-azacitidine,azacitidinum inn-latin PubChem CID: 9444 ChEBI: CHEBI:2038 IUPAC Name: 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydro-1,3,5-triazin-2-one SMILES: NC1=NC(=O)N(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O