CAS RN 4393-09-3
CAS RN 4393-09-3
IUPAC Name:
1-(2,3-dimethoxyphenyl)methanamine
Synonyms:
1-(2,3-dimethoxyphenyl)methanamine
Molecular Weight (g/mol):
167.21
Molecular Formula:
C9H13NO2
InChi Key:
LVMPWFJVYMXSNY-UHFFFAOYSA-N
SMILES:
COC1=CC=CC(CN)=C1OC
1
–
2
of
2
results
2
Thermo Scientific Acros 2,3-Dimethoxybenzylamine, 99%, Thermo Scientific™
CAS: 4393-09-3 Molecular Formula: C9H13NO2 Molecular Weight (g/mol): 167.21 MDL Number: MFCD00052392 InChI Key: LVMPWFJVYMXSNY-UHFFFAOYSA-N