CAS RN 48212-99-3
CAS RN 48212-99-3
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
N4-Anisoyl-2'-deoxycytidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N4-Anisoyl-2'-deoxycytidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N4-Anisoyl-2'-deoxycytidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.