accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 516-54-1

H3CO(S)(S)(R)(S)(R)OH(S)H3C(S)(S)H3C

CAS RN 516-54-1

IUPAC Name: 1-[(1S,3aS,3bR,5aS,7R,9aS,9bS,11aS)-7-hydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]ethan-1-one
Synonyms: allopregnanolone 3a-hydroxy-5a-pregnan-20-one 3-α,5-α-pregnanolone 3a,5a-pregnanolone 3-α-OH dhp 3a-hydroxy-5a-pregnane-20-one 5a-pregnan-3a-ol-20-one 3a,5a-tetrahydroprogesterone 5a-pregnane-3a-ol-20-one allopregnan-3α-ol-20-one
Molecular Weight (g/mol): 318.5
Molecular Formula: C21H34O2
InChi Key: AURFZBICLPNKBZ-SYBPFIFISA-N
SMILES: CC(=O)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products