accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 5534-09-8

H3COOO(R)OOCH3(S)H3C(S)(S)O(S)H3C(R)Cl(S)OH(S)H3C

CAS RN 5534-09-8

IUPAC Name: 2-[(1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-chloro-10-hydroxy-2,9a,11a-trimethyl-7-oxo-1-(propanoyloxy)-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl propanoate
Synonyms: beconase Spir beclovent beclomethasone dipropionate andion beclate vancenase vanceril becotide beclometasone dipropionate
Molecular Weight (g/mol): 521.05
Molecular Formula: C28H37ClO7
InChi Key: KUVIULQEHSCUHY-XYWKZLDCSA-N
SMILES: CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products