accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 59084-06-9

ONONH2N

CAS RN 59084-06-9

IUPAC Name: 4-(2-nitrophenyl)piperazin-1-ium
Synonyms: 4-(2-nitrophenyl)piperazin-1-ium
Molecular Weight (g/mol): 208.24
Molecular Formula: C10H14N3O2
InChi Key: YJRCDSXLKPERNV-UHFFFAOYSA-O
SMILES: [O-][N+](=O)C1=CC=CC=C1N1CC[NH2+]CC1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products