accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 70539-42-3

OOONOOOOOOONOO

CAS RN 70539-42-3

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 1-[2-({4-[(2,5-dioxopyrrolidin-1-yl)oxy]-4-oxobutanoyl}oxy)ethyl] butanedioate
Synonyms: 2,5-dioxopyrrolidin-1-yl 1-[2-({4-[(2,5-dioxopyrrolidin-1-yl)oxy]-4-oxobutanoyl}oxy)ethyl] butanedioate
Molecular Weight (g/mol): 456.36
Molecular Formula: C18H20N2O12
InChi Key: QLHLYJHNOCILIT-UHFFFAOYSA-N
SMILES: O=C(CCC(=O)ON1C(=O)CCC1=O)OCCOC(=O)CCC(=O)ON1C(=O)CCC1=O
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products