accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 75428-45-4

ONOSO

CAS RN 75428-45-4

IUPAC Name: 5-nitrothiophene-3-carbaldehyde
Synonyms: 5-nitrothiophene-3-carbaldehyde
Molecular Weight (g/mol): 157.14
Molecular Formula: C5H3NO3S
InChi Key: JYVNBTUBCTXNBI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(C=O)=CS1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products