CAS RN 818-23-5
CAS RN 818-23-5
IUPAC Name:
pentadecan-8-one
Synonyms:
8-pentadecanone
Molecular Weight (g/mol):
226.4
Molecular Formula:
C15H30O
InChi Key:
PQYGSSYFJIJDFK-UHFFFAOYSA-N
SMILES:
CCCCCCCC(=O)CCCCCCC