CAS RN 6190-65-4
IUPAC Namn:
6-chloro-N2-(propan-2-yl)-1,3,5-triazine-2,4-diamine
Synonymer:
desethylatrazine deethylatrazine desethyl atrazine 4-deethylatrazine deethylatrazin des-ethyl atrazine C6H10ClN5
Molekylvikt (g/mol):
187.63
Molekylär sammansättning:
C6H10ClN5
InChi Key:
DFWFIQKMSFGDCQ-UHFFFAOYSA-N
SMILES:
CC(C)NC1=NC(N)=NC(Cl)=N1
1
–
4
av
4
Resultat
1
2-Amino-4-isopropylamino-6-chlorotriazine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Amino-4-isopropylamino-6-chlorotriazine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Amino-4-isopropylamino-6-chlorotriazine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.