missing translation for 'onlineSavingsMsg'
Learn More
2-Acetylfuran, 99% Product Code.: 10796121

2-Acetylfuran, 99%

Product Code. p-49506 Shop All Thermo Scientific Chemicals Products
100 g, Glass bottle, 100g
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass bottle
Unit Size:
100g
250g
This item is not returnable. View return policy

Product Code. 10796121

Brand: Thermo Scientific Acros 102551000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1192-62-7 | C6H6O2 | 110.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1192-62-7
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.11
MDL Number MFCD00003242
InChI Key IEMMBWWQXVXBEU-UHFFFAOYSA-N
Synonym 2-acetylfuran, acetylfuran, 1-furan-2-yl ethanone, 2-furyl methyl ketone, methyl 2-furyl ketone, 1-2-furyl ethanone, 2-furylethanone, 2-acetyl furan, ketone, 2-furyl methyl, ethanone, 1-2-furanylShow MoreShow Less
PubChem CID 14505
ChEBI CHEBI:59983
IUPAC Name 1-(furan-2-yl)ethanone
SMILES CC(=O)C1=CC=CO1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 29.0°C to 30.0°C
Color Yellow to Brown
Density 1.0980g/mL
Boiling Point 67.0°C (10.0 mmHg)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 0.2% max. 2,5-Diacetyl furan (GC)
Packaging Glass bottle
Refractive Index 1.5060 to 1.5080
Quantity 100 g
Beilstein 17, 286
Specific Gravity 1.098
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether
Formula Weight 110.11
Percent Purity 99%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2-Acetylfuran, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0470466 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code102551000,102552500
Lot NumberA0456828 Certificate TypeCertificate of Analysis Date10/10/2023 Product Code102551000,102552500
Lot NumberA0450654 Certificate TypeCertificate of Analysis Date02/05/2023 Product Code102551000,102552500
Lot NumberA0393967 Certificate TypeCertificate of Analysis Date12/12/2022 Product Code102551000,102552500
Lot NumberA0439677 Certificate TypeCertificate of Analysis Date22/03/2022 Product Code102551000,102552500
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H311-Toxic in contact with skin.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Acetylfuran, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.