Learn More
Wheaton™ 60 mL Polypropylene Sample Container
Sterile sample container offers sample security and user safety with leak free performance.
1505.00 SEK
Specifications
Product Type | Specimen Container |
---|---|
Closure Included | Yes |
Sterility | Sterile |
Capacity (Metric) | 60 mL |
Certifications/Compliance | EU Directive 98/79/EC |
Product Code | Brand | Label | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Label | Price | Quantity & Availability | |||||
16474969
|
Wheaton™
360104 |
No Label |
1505.00 SEK
Pack of 300 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16464969
|
Wheaton™
360103PP |
Printed |
1505.00 SEK
Pack of 300 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- For general sample collection, transport, and storage
- Wide container range to suit most user needs
Specifications
Specimen Container | |
Sterile | |
EU Directive 98/79/EC | |
Polyethylene | |
44.2 mm | |
300/Cs. |
Yes | |
60 mL | |
White | |
Screw Cap | |
60.2 mm | |
50/Pk. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Wheaton™ 60 mL Polypropylene Sample Container