Learn More
Alpha SNAP Mouse anti-Human, Unconjugated, Clone: 1E2A2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67323-1-IG-20UL
 View more versions of this product
Product Code. 16889885
  • 1645.54 SEK / 20µL
Description
The Alpha SNAP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Alpha SNAP. This antibody recognizes human, pig, mouse, rat antigen. The Alpha SNAP antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
Alpha SNAP
Monoclonal
1000 ÎĽg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1E2A2
Unconjugated
20 ÎĽL
8775
IgG2b
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Alpha SNAP Mouse anti-Human, Unconjugated, Clone: 1E2A2, Proteintech > 20 ÎĽL