Learn More
Ansell AlphaTec™ 2000 STANDARD Trousers Bound
Comfortable trousers featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress.
5575.00 SEK
Specifications
Material | Microporous Non-woven Polyethylene Laminate |
---|---|
Color | White |
Seam Type | Bound |
Certifications/Compliance (Europe) | Partial Body Protection Only, EN 13034:2005 + A1:2009, EN 14126:2003, EN ISO 13688:2013 |
Style | W20B301 |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
16874790
|
Ansell
WH20B-00301-05 |
X-Large |
5575.00 SEK
Pack of 80 |
Estimated Shipment: 24-10-2025
Log in to see stock. |
||||||
16884790
|
Ansell
WH20B-00301-06 |
2X-Large |
5575.00 SEK
Pack of 80 |
Estimated Shipment: 24-10-2025
Log in to see stock. |
||||||
Description
- Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
- Optimized fit, improving wearer comfort and safety
- Low Linting
Specifications
Microporous Non-woven Polyethylene Laminate | |
Bound | |
W20B301 |
White | |
Partial Body Protection Only, EN 13034:2005 + A1:2009, EN 14126:2003, EN ISO 13688:2013 | |
Clean Room Cleaning and Preparing, Production Line Support & Maintenance, Maintenance of Plant and Machines, Veterinary Services |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Ansell AlphaTec™ 2000 STANDARD Trousers Bound