Learn More
Ansell AlphaTec™ 2500 STANDARD
Chemical protective suits with durable and breathable microporous laminate technology and Type 4/5/6 protection.
5735.00 SEK - 8595.00 SEK
Specifications
Color | White |
---|---|
For Use With (Application) | Transferring Liquids and Solids, Unexpected Leakages Spills or Other Releases, Plant Disinfection and Sanitization, Tank Cleaning, Emergency Response, Spill or Leakage Cleanup, Maintenance, Contaminated Environments |
Material | Microporous Non-woven Polypropylene Laminate |
Seam Type | Welded |
Style | W25W122 |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
16844860
|
Ansell
WH25W-00122-02 |
Small |
8595.00 SEK
Pack of 30 |
Estimated Shipment: 29-08-2025
Log in to see stock. |
||||||
16854860
|
Ansell
WH25W-00122-03 |
Medium |
8595.00 SEK
Pack of 30 |
Estimated Shipment: 29-08-2025
Log in to see stock. |
||||||
16864860
|
Ansell
WH25W-00122-04 |
Large |
8595.00 SEK
Pack of 30 |
Estimated Shipment: 29-08-2025
Log in to see stock. |
||||||
16874860
|
Ansell
WH25W-00122-05 |
X-Large |
8595.00 SEK
Pack of 30 |
Estimated Shipment: 29-08-2025
Log in to see stock. |
||||||
16884860
|
Ansell
WH25W-00122-06 |
2X-Large |
7170.00 SEK
Pack of 25 |
Estimated Shipment: 29-08-2025
Log in to see stock. |
||||||
16894860
|
Ansell
WH25W-00122-07 |
3X-Large |
7170.00 SEK
Pack of 25 |
Estimated Shipment: 29-08-2025
Log in to see stock. |
||||||
16804870
|
Ansell
WH25W-00122-08 |
4X-Large |
5735.00 SEK
Pack of 20 |
Estimated Shipment: 29-08-2025
Log in to see stock. |
||||||
Description
- Air and water vapour permeable (breathable) fabric helps reduce the risk of heat stress
- Ultra low linting to help reduce the risk of cross contamination in critical areas
- Achieves the highest classifications for protection from biological agents in accordance with EN 14126 and ASTM F 1671 for penetration of blood, body fluids and blood-borne pathogens
- Anti-static
Specifications
White | |
Microporous Non-woven Polypropylene Laminate | |
W25W122 |
Transferring Liquids and Solids, Unexpected Leakages Spills or Other Releases, Plant Disinfection and Sanitization, Tank Cleaning, Emergency Response, Spill or Leakage Cleanup, Maintenance, Contaminated Environments | |
Welded | |
65 gsm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Ansell AlphaTec™ 2500 STANDARD