Learn More
Ansell AlphaTec™ 3000 Ultrasonically Welded Model 103
AlphaTec™ 3000 Ultrasonically Welded Model 103 features a durable, comfortable multi-layer chemical barrier with dual-zip systems and double cuffs.
490.00 SEK
Specifications
Product Type | Chemical Protective Clothing |
---|---|
Gender | Unisex |
Material | Multi-layer Barrier Laminate Fabric |
Color | Yellow |
Disposable | Reusable |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17999140
|
Ansell
YE30W-00103-07 |
3X-Large |
490.00 SEK
Each |
Estimated Shipment: 18-12-2025
Log in to see stock. |
||||||
17989150
|
Ansell
YE30W-00103-08 |
4X-Large |
490.00 SEK
Each |
Estimated Shipment: 18-12-2025
Log in to see stock. |
||||||
17929150
|
Ansell
YE30W-00103-09 |
5X-Large |
490.00 SEK
Each |
Estimated Shipment: 18-12-2025
Log in to see stock. |
||||||
Description
- Reinforced protection: The AlphaTec™ 3000 Ultrasonically Welded - Model 103 protective suit features multi-layered fabric that ensures an effective barrier against numerous inorganic chemicals and biohazards
- Enhanced features: This protective barrier is further reinforced by its double cuffs, dual-zip systems and double zip enclosure, which extends wear life
- Increased visibility: Thanks to its highly visible bright yellow color, this protective outer garment remains particularly noticeable in low-lit environments, enhancing wearer and workplace safety
- Assured antistatic properties: In addition, it complies with EN 1149-5 antistatic standards
Specifications
Chemical Protective Clothing | |
Multi-layer Barrier Laminate Fabric | |
Reusable | |
Elastic | |
Collared | |
Double Zipper | |
Non-sterile | |
Liquid, Chemicals, Particle, Anti-static, Bloodborne Pathogens | |
EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, CE Number, EN 1073-2:2002, EN 1149-5:2018, EN ISO 13688:2013, EN 14126:2003 | |
None | |
3000 103 |
Unisex | |
Yellow | |
Elastic | |
Elastic, Double | |
Unhooded | |
Welded | |
Type 3-B, Type 4-B, Type 5-B, Type 6-B | |
Coverall with Collar, Double Zip Closure, Double Cuffs, Elasticated Waist, Double Cuffs and Ankles | |
No | |
Transferring liquids and solids between vessels and tanks and process equipment, Unexpected leakages spills or other releases, Opening and draining pumps valves or lines, Loading and unloading trucks and vehicles, Filling, blending and charging of raw materials, Product inspection, Mixing or loading chemicals, Cleaning of plant and machines |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Ansell AlphaTec™ 3000 Ultrasonically Welded Model 103