missing translation for 'onlineSavingsMsg'
Learn More
Invitrogen™ Human CD40 Ligand (TNFSF5) Recombinant Protein, eBioscience™ Product Code.: 15558836

Invitrogen™ Human CD40 Ligand (TNFSF5) Recombinant Protein, eBioscience™

Product Code. 15558836
50 μg, 50µg
Click to view available options
Quantity:
10 μg
50 μg
500 μg
Unit Size:
10µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 15558836

Brand: Invitrogen™ 34890281

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

CD40 Ligand (CD40-L), or CD154, is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD40 Ligand stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD40 Ligand has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. It also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma, TNF-alpha, and IL-2. Description: CD40 ligand, (CD40L, also known as CD154, TRAP or gp39) is a membrane glycoprotein expressed on activated CD4+ T-cells, NK cells, mast cells, basophils and eosinophils. The CD40-CD40L interaction stimulates B cell immune response which includes cell surface antigen expression, cell cycle activation, Ig isotype switching, Ig secretion and memory generation. The CD40-CD40L interaction also plays important roles in monocyte and dendritic cell activation, T-cell co-stimulation and cytokine production. It has been reported that the CD40-CD40L interaction is involved in the pathogenesis of amyloid pathology in Alzheimer disease. Applications Reported: This Human CD154 (CD40 Ligand) Recombinant Protein is biologically active and can induce IL-8 secretion by normal human peripheral blood cells. Applications Tested: The activity of this protein was determined by measuring IL-8 production by normal human peripheral blood cells. Source: E. coli (amino acids Gln114-Leu261; accession # NP_000065.1). Bioactivity: The ED50 of this protein, as measured by stimulation of IL-8 production by normal human peripheral blood cells, is less than or equal to 5-10ng/mL. Endotoxin: Less than 0.1ng/ug cytokine as determined by the LAL assay. Purity: >98% as determined by SDS-PAGE and HPLC. Molecular Weight: 16.3 kDa. Storage and handling: For best recovery, quick spin vial prior to opening. Use in sterile environment. Reconstitute in 5-10 mM sodium phosphate, pH 7.5 to a concentration of 0.1-1mg/mL. Do not vortex. Store lyophilized protein at less than or equal to -20°C. Reconstituted protein solution can be stored with carrier protein (e.g., 0.1% BSA) at less than or equal to -20°C.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P29965
For Use With (Application) Control, Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 959
Molecular Weight (g/mol) 16.3kDa
Name Human CD154 (CD40 Ligand) Carrier-Free
Purification Method SDS-PAGE and HPLC
Quantity 50 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Structural Form E. coli (amino acids Gln114-Leu261; Accession # NP_000065.1)
Endotoxin Concentration Less than 0.1ng/μg cytokine as determined by the LAL assay
Common Name Human CD154 (CD40 Ligand) Carrier-Free
Gene Symbol CD40LG
Biological Activity The ED50 of this protein, as measured by stimulation of IL-8 production by normal human peripheral blood cells, is less than or equal to 5-10ng/mL.
Product Type Recombinant Protein, Carrier-Free
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade Affinity chromatography
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Human CD40 Ligand (TNFSF5) Recombinant Protein, eBioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.