missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASBT Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25245-1-AP-20UL
This item is not returnable.
View return policy
Specifications
ASBT | |
Polyclonal | |
Unconjugated | |
SLC10A2 | |
ASBT, IBAT, ISBT, NTCP2, SLC10A2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20494, 6555 | |
-20°C | |
Liquid |
Immunoprecipitation | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P70172, Q12908 | |
SLC10A2 | |
ASBT Fusion Protein Ag18808 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ASBT Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction