Learn More
Hecht Karl™ Assistent™ Borosilicate Weighing Bottles
192.00 SEK - 590.00 SEK
Specifications
Includes | Interchangeable NS cover and frosted label |
---|---|
Material | Borosilicate Glass |
Product Type | Weighing Bottle |
Closure Type | NS Cover |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10639831
|
Hecht Karl™
44250001 |
10 mL |
226.00 SEK
Each |
Estimated Shipment: 20-08-2025
Log in to see stock. |
||||||
10309031
|
Hecht Karl™
44251011 |
15 mL |
255.00 SEK
Each |
Estimated Shipment: 15-08-2025
Log in to see stock. |
||||||
10044071
|
Hecht Karl™
44251013 |
30 mL |
192.00 SEK
Each |
Estimated Shipment: 19-08-2025
Log in to see stock. |
||||||
10630792
|
Hecht Karl™
44251014 |
45 mL |
280.00 SEK
Each |
Estimated Shipment: 19-08-2025
Log in to see stock. |
||||||
10388881
|
Hecht Karl™
44250005 |
70 mL |
242.00 SEK
Each |
Estimated Shipment: 22-09-2025
Log in to see stock. |
||||||
10238011
|
Hecht Karl™
44251016 |
80 mL |
590.00 SEK
Each |
Estimated Shipment: 20-08-2025
Log in to see stock. |
||||||
10185391
|
Hecht Karl™
44250006 |
110 mL |
377.00 SEK
Each |
Estimated Shipment: 20-08-2025
Log in to see stock. |
||||||
Specifications
Interchangeable NS cover and frosted label | |
Borosilicate Glass | |
Weighing Bottle | |
NS Cover |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Hecht Karl™ Assistent™ Borosilicate Weighing Bottles