missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDH2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27207-1-AP-20UL
This item is not returnable.
View return policy
Description
The function of this protein remains unknown.Specifications
BDH2 | |
Polyclonal | |
Unconjugated | |
BDH2 | |
BDH2, DHRS6, EFA6R, Oxidoreductase UCPA, PRO20933, SDR15C1, UCPA OR, UNQ6308 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56898 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BUT1 | |
BDH2 | |
BDH2 Fusion Protein Ag24934 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BDH2 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction