Learn More
Bohlender™ BOLA™ PTFE/PPS HT Replacement Inner Parts
Consisting of a v-ring (PPS), a tapered ring and a sealing ring (both PTFE) as well as an o-ring (FKM) for tubing diameters under 3mm (not exposed to the medium).
264.00 SEK - 1460.00 SEK
Specifications
Certifications/Compliance | FDA |
---|---|
Autoclavable | Autoclavable |
Material | PPS, PTFE |
Min. Operating Temperature (Metric) | -200°C |
Color | White |
Product Code | Brand | Thread Size | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Tubing Size | Price | Quantity & Availability | |||||
10409391
|
Bohlender™
D639-22 |
GL 18 | 2mm O.D. |
264.00 SEK
Each |
Estimated Shipment: 30-07-2025
Log in to see stock. |
|||||
10616683
|
Bohlender™
D640-42 |
GL 25 | (1/8 in.) 3.2mm O.D. |
534.00 SEK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
|||||
10043371
|
Bohlender™
D640-18 |
GL 25 | (1/16 in.) 1.6mm O.D. |
327.00 SEK
Each |
Estimated Shipment: 27-08-2025
Log in to see stock. |
|||||
10267451
|
Bohlender™
D642-74 |
GL 45 | (1 in.) 25.4mm O.D. |
1210.00 SEK
Each |
Estimated Shipment: 22-08-2025
Log in to see stock. |
|||||
10651522
|
Bohlender™
D640-46 |
GL 25 | 4mm O.D. |
322.00 SEK
Each |
Estimated Shipment: 22-08-2025
Log in to see stock. |
|||||
10134921
|
Bohlender™
D641-38 |
GL 32 | 6mm O.D. |
1105.00 SEK
Each |
Estimated Shipment: 27-08-2025
Log in to see stock. |
|||||
10491872
|
Bohlender™
D640-22 |
GL 25 | 2mm O.D. |
323.00 SEK
Each |
Estimated Shipment: 06-08-2025
Log in to see stock. |
|||||
10368941
|
Bohlender™
D641-60 |
GL 32 | 12mm O.D. |
1245.00 SEK
Each |
Estimated Shipment: 27-08-2025
Log in to see stock. |
|||||
10124971
|
Bohlender™
D641-56 |
GL 32 | 10mm O.D. |
1030.00 SEK
Each |
Estimated Shipment: 22-08-2025
Log in to see stock. |
|||||
10504212
|
Bohlender™
D641-42 |
GL 32 | (1/4 in.) 6.35mm O.D. |
1025.00 SEK
Each |
Estimated Shipment: 22-08-2025
Log in to see stock. |
|||||
10449191
|
Bohlender™
D641-78 |
GL 32 | 16mm O.D. |
1025.00 SEK
Each |
Estimated Shipment: 22-08-2025
Log in to see stock. |
|||||
10575742
|
Bohlender™
D642-44 |
GL 45 | 12mm O.D. |
1460.00 SEK
Each |
Estimated Shipment: 22-08-2025
Log in to see stock. |
|||||
10184631
|
Bohlender™
D642-76 |
GL 45 | 26mm O.D. |
999.00 SEK
Each |
Estimated Shipment: 22-08-2025
Log in to see stock. |
Vous avez déjà un compte ? Connectez-vous pour voir vos prix et disponibilités. Nouveau ici ? Inscrivez-vous dès aujourd'hui ! |
||||
10144921
|
Bohlender™
D641-46 |
GL 32 | 8mm O.D. |
1045.00 SEK
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
Vous avez déjà un compte ? Connectez-vous pour voir vos prix et disponibilités. Nouveau ici ? Inscrivez-vous dès aujourd'hui ! |
||||
Description
- Medium is only exposed to PTFE
- Good chemical resistance
- Withstand pressure up to 10 bar
- Vacuum suitable
Spécification
FDA | |
PPS, PTFE | |
White | |
250°C |
Autoclavable | |
-200°C | |
10 bar |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.