Learn More
Lenz Laborglasinstrumente™ Separating Funnels Acc. to Squibb, with PTFE Plug
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels Acc. to Squibb, with PTFE Plug are manufactured from durable DURAN™ borosilicate glass.
789.00 SEK - 1875.00 SEK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10078420
|
Lenz Laborglasinstrumente™
04014170 |
1000 mL |
1410.00 SEK
Pack of 2 |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||||
10424111
|
Lenz Laborglasinstrumente™
04014173 |
2000 mL |
1544.40 SEK
Each |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
10567541
|
Lenz Laborglasinstrumente™
04014137 |
100 mL |
789.00 SEK
Pack of 2 |
Estimated Shipment: 30-06-2025
Log in to see stock. |
||||||
10633161
|
Lenz Laborglasinstrumente™
04014149 |
250 mL |
963.00 SEK
Pack of 2 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
10415431
|
Lenz Laborglasinstrumente™
04014158 |
500 mL |
1875.00 SEK
Pack of 2 |
In Stock
Log in to see stock. |
||||||
10567351
|
Lenz Laborglasinstrumente™
04014128 |
50 mL |
830.00 SEK
Pack of 2 |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||||
Description
- Without scale
- With NS socket and PE stopper
- With screwthread retaining device
- Corrosion-free and resistant to thermal shock
Specifications
Borosilicate Glass | |
Polyethylene |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Separating Funnels Acc. to Squibb, with PTFE Plug