missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution Ammonia-Ammonium Chloride, Pure, for Water Hardness Testing, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
Description
Buffer Solution Ammonia-Ammonium Chloride, Pure, for Water Hardness Testing

Specifications
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 2.5 L |
Packaging | HDPE plastic bottle |
Vapor Pressure | 330mmHg at 20°C |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Safety and Handling



Product Identifier
- Buffer solution ammonia-ammonium chloride
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/A§.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buffer Solution Ammonia-Ammonium Chloride, Pure, for Water Hardness Testing, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction