missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C17orf28 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21174-1-AP-150UL
This item is not returnable.
View return policy
Description
CQ028 may play an important role in the development of cancers in a broad range of tissues.Specifications
C17orf28 | |
Polyclonal | |
Unconjugated | |
HID1 | |
C17orf28, DMC1, HID1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
217310, 283987, 287822 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8IV36, Q8R1F6 | |
HID1 | |
C17orf28 Fusion Protein Ag15427 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C17orf28 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction