missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf103 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26115-1-AP-20UL
This item is not returnable.
View return policy
Description
The specific function of this protein remains unknown.Specifications
C1orf103 | |
Polyclonal | |
Unconjugated | |
LRIF1 | |
C1orf103, LRIF1, Receptor interacting factor 1, RIF1, RP11 96K19.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
321000, 55791 | |
-20°C | |
Liquid |
Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5T3J3, Q8CDD9 | |
Lrif1 | |
C1orf103 Fusion Protein Ag23494 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C1orf103 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction