Learn More
MedChemExpress Calcein tetraethyl ester
MedChemExpress Calcein tetraethyl ester is a fluorescent dye, is used in biology as it can be transported through the cellular membrane into live cells, which makes it useful for testing of cell viability and for short-term labeling of cells.

1540.50 SEK - 8356.04 SEK
Specifications
Content And Storage | 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
---|---|
Product Type | Dye Reagents |
Shipping Condition | Room temperature |
Solubility | DMSO : 25 mg/mL (34.03 mM; Need ultrasonic) ∣ H2O : < 0.1 mg/mL (insoluble) |
Target | HBV |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16488906
![]() |
MedChemExpress
HY-D0830 |
100 mg |
8356.04 SEK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16438768
![]() |
MedChemExpress
HY-D0830 |
10 mg |
1540.50 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16414687
![]() |
MedChemExpress
HY-D0830 |
50 mg |
4637.06 SEK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) | |
Room temperature | |
HBV | |
1170856-93-5 |
Dye Reagents | |
DMSO : 25 mg/mL (34.03 mM; Need ultrasonic) ∣ H2O : < 0.1 mg/mL (insoluble) | |
734.75 g/mol | |
Fluorexon tetraethyl ester |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.