Learn More
Lenz Laborglasintrumente™ Chromatographic Columns with Frit
Chromatographic columns with frit, NS socket and NS cone, and with PTFE stopcock
1550.00 SEK - 1736.80 SEK
Specifications
Quantity 1
Length (Metric) 400 mm
Material DURAN™ Tubing
View More Specs
Products 2
Product Code Brand Column Capacity Price Quantity & Availability  
Product Code Brand Column Capacity Price Quantity & Availability  
10667970
View Documents
Lenz Laborglasintrumente™
05450902
120 mL
1550.00 SEK
Each
Only null left
Estimated Shipment: 28-07-2025
Add to basket
11914021
View Documents
Lenz Laborglasintrumente™
05450904
280 mL
1736.80 SEK
Each
Only null left
Estimated Shipment: 28-07-2025
Add to basket
Description
Description
  • With sintered frit (porosity: 0)
  • With suction tube (thread GL 18)
  • Made of DURAN™ tubing
Specifications
Specifications
1
400 mm
DURAN™ Tubing
Documents
Product Certifications
Product Content Correction
Product Title
Lenz Laborglasintrumente™ Chromatographic Columns with Frit