Learn More
Fisherbrand™ Combs for MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit, 1mm Thick
For use in preparatory electrophoresis and high-throughput screening.
277.00 SEK - 312.00 SEK
Specifications
For Use With (Equipment) | MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit |
---|---|
Type | Comb |
Thickness (Metric) | 1 mm |
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | |||||
11843363
|
Fisherbrand™
MS15-10-1 |
10 | 45 ÎĽL |
312.00 SEK
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
11883363
|
Fisherbrand™
MS15-10MC-1 |
10 | 29 ÎĽL |
312.00 SEK
Each |
Estimated Shipment: 06-08-2025
Log in to see stock. |
|||||
11813373
|
Fisherbrand™
MS15-1-1 |
1 Prep, 1 Marker | 495 ÎĽL |
312.00 SEK
Each |
Estimated Shipment: 20-08-2025
Log in to see stock. |
|||||
11853373
|
Fisherbrand™
MS15-1/2-1 |
12 | 41 ÎĽL | N/A | ||||||
11893373
|
Fisherbrand™
MS15-14MC-1 |
14 | 29 ÎĽL |
312.00 SEK
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
11833383
|
Fisherbrand™
MS15-16MC-1 |
16 | 27 ÎĽL |
312.00 SEK
Each |
Estimated Shipment: 24-07-2025
Log in to see stock. |
|||||
11873383
|
Fisherbrand™
MS15-18MC-1 |
18 | 11 ÎĽL | N/A | ||||||
11823393
|
Fisherbrand™
MS15-20-1 |
20 | 21 ÎĽL |
302.00 SEK
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
11853393
|
Fisherbrand™
MS15-2-1 |
2 Prep, 2 Marker | 225 ÎĽL |
312.00 SEK
Each |
Estimated Shipment: 20-08-2025
Log in to see stock. |
|||||
11833403
|
Fisherbrand™
MS15-30MC-1 |
30 | 13 ÎĽL | N/A | ||||||
11873403
|
Fisherbrand™
MS15-35-1 |
35 | 10 ÎĽL |
312.00 SEK
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
11813413
|
Fisherbrand™
MS15-4-1 |
4 Prep, 2 Marker | 122 ÎĽL |
312.00 SEK
Each |
Estimated Shipment: 12-08-2025
Log in to see stock. |
|||||
Description
- Supplied as standard
- Recommended for high resolution gels and publication quality data
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 10, 14, 16, 18, 28 and 30-sample combs are Multichannel pipettor compatible
Specifications
MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit | |
Comb | |
1 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Combs for MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit, 1mm Thick