missing translation for 'onlineSavingsMsg'
Learn More
Copper(II) picolinate, 98%, Thermo Scientific™ Product Code.: 15575946

Copper(II) picolinate, 98%, Thermo Scientific™

Product Code. 15575946 Shop All Thermo Scientific Chemicals Products
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15575946

Brand: Thermo Scientific Acros 457200050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 14050-01-2
Molecular Formula C12H8CuN2O4
Molecular Weight (g/mol) 307.75
InChI Key GNWVUSJJFONIKF-UHFFFAOYSA-L
Synonym copper ii picolinate, unii-96e596pypp, copper picolinate, copper, bis picolinato, copper picolinate anhydrous, copper, bis 2-pyridinecarboxylato-kappan1,kappao2, copper 2+ ion dipicolinate, copper,bis 2-pyridinecarboxylato-kn1,ko2-, hydrate 1:2Show MoreShow Less
PubChem CID 14963587
IUPAC Name copper;pyridine-2-carboxylate
SMILES C1=CC=NC(=C1)C(=O)[O-].C1=CC=NC(=C1)C(=O)[O-].[Cu+2]

Specifications

Quantity 5 g
Assay Percent Range 97.5% min. (Iodometry)
Formula Weight 307.75
Percent Purity 98%
Infrared Spectrum Authentic
Loss on Drying 0.02
Chemical Name or Material Copper(II) picolinate
compliance-icons compliance-icons
Product Identifier
  • Copper(II) picolinate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper(II) picolinate, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.