missing translation for 'onlineSavingsMsg'
Learn More
Learn More
α-Cyclodextrin, MP Biomedicals™
339.00 SEK - 4075.00 SEK
Chemical Identifiers
CAS | 10016-20-3 |
---|---|
Molecular Formula | C36H60O30 |
Molecular Weight (g/mol) | 972.846 |
InChI Key | HFHDHCJBZVLPGP-UHFFFAOYSA-N |
Synonym | cyclohexaamylose, cyclomaltohexose, .alpha.-cyclodextrin, schardinger alpha-dextrin, .alpha.-dextrin, .alpha.-cycloamylose, bdbm4, dextrin, .alpha.-cyclo, .alpha.-schardinger dextrin |
PubChem CID | 320760 |
SMILES | C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11454502
|
MP Biomedicals
0219005201 |
1 g |
339.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11464502
|
MP Biomedicals
0219005205 |
5 g |
908.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11474502
|
MP Biomedicals
0219005225 |
25 g |
4075.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Chemische Identifikatoren
10016-20-3 | |
972.846 | |
cyclohexaamylose, cyclomaltohexose, .alpha.-cyclodextrin, schardinger alpha-dextrin, .alpha.-dextrin, .alpha.-cycloamylose, bdbm4, dextrin, .alpha.-cyclo, .alpha.-schardinger dextrin | |
C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)O)O |
C36H60O30 | |
HFHDHCJBZVLPGP-UHFFFAOYSA-N | |
320760 |
Spezifikation
10016-20-3 | |
C36H60O30 | |
HFHDHCJBZVLPGP-UHFFFAOYSA-N | |
972.846 | |
972.9 | |
α-Cyclodextrin |
White | |
cyclohexaamylose, cyclomaltohexose, .alpha.-cyclodextrin, schardinger alpha-dextrin, .alpha.-dextrin, .alpha.-cycloamylose, bdbm4, dextrin, .alpha.-cyclo, .alpha.-schardinger dextrin | |
C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)O)O | |
320760 | |
Powder |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfĂĽllen.
Name des Produkts
α-Cyclodextrin, MP Biomedicals™