missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-alpha-Tocopherol acetate powder, MP Biomedicals™
641.00 SEK - 6265.00 SEK
Chemical Identifiers
CAS | 7695-91-2 |
---|---|
Molecular Formula | C31H52O3 |
Molecular Weight (g/mol) | 472.754 |
MDL Number | MFCD00072042 |
InChI Key | ZAKOWWREFLAJOT-CEFNRUSXSA-N |
Synonym | vitamin e acetate, tocopherol acetate, alpha-tocopherol acetate, alfacol, ecofrol, d-alpha-tocopherol acetate, contopheron, tofaxin, ephynal acetate, econ |
PubChem CID | 86472 |
ChEBI | CHEBI:32321 |
IUPAC Name | [(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate |
SMILES | CC1=C2C(=C(C(=C1C)OC(=O)C)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11427630
|
MP Biomedicals
0210055525 |
25 g |
641.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11437630
|
MP Biomedicals
0210055580 |
100 g |
1890.00 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11447630
|
MP Biomedicals
0210055590 |
500 g |
6265.00 SEK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
7695-91-2 | |
472.754 | |
ZAKOWWREFLAJOT-CEFNRUSXSA-N | |
86472 | |
[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate |
C31H52O3 | |
MFCD00072042 | |
vitamin e acetate, tocopherol acetate, alpha-tocopherol acetate, alfacol, ecofrol, d-alpha-tocopherol acetate, contopheron, tofaxin, ephynal acetate, econ | |
CHEBI:32321 | |
CC1=C2C(=C(C(=C1C)OC(=O)C)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C |
Specifications
7695-91-2 | |
250U/g | |
MFCD00072042 | |
ZAKOWWREFLAJOT-CEFNRUSXSA-N | |
[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate | |
86472 | |
Powder |
0.96g/mL at 20°C | |
C31H52O3 | |
vitamin e acetate, tocopherol acetate, alpha-tocopherol acetate, alfacol, ecofrol, d-alpha-tocopherol acetate, contopheron, tofaxin, ephynal acetate, econ | |
CC1=C2C(=C(C(=C1C)OC(=O)C)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C | |
472.754 | |
CHEBI:32321 | |
DL-α-Tocopherol acetate, Cell Biology and Analysis |
Safety and Handling
missing translation for 'einecsNumber' : 231-710-0
Recommended Storage : Store at 4°C.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DL-alpha-Tocopherol acetate powder, MP Biomedicals™