missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doxycycline hydrochloride, MP Biomedicals™
677.00 SEK - 1650.00 SEK
Chemical Identifiers
CAS | 10592-13-9 |
---|---|
Molecular Formula | C22H25ClN2O8 |
Molecular Weight (g/mol) | 480.90 |
MDL Number | MFCD03427564 |
InChI Key | VLUQVUWDECWBTL-UQVCFKGQSA-N |
Synonym | Biocamycin, Tetradox |
PubChem CID | 54706018 |
SMILES | [Cl-].C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H]([NH+](C)C)C(=O)\C(=C(/N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11460892
|
MP Biomedicals
0219504401 |
1 g |
677.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470892
|
MP Biomedicals
0219504405 |
5 g |
1650.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
10592-13-9 | |
480.90 | |
VLUQVUWDECWBTL-UQVCFKGQSA-N | |
54706018 |
C22H25ClN2O8 | |
MFCD03427564 | |
Biocamycin, Tetradox | |
[Cl-].C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H]([NH+](C)C)C(=O)\C(=C(/N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12Show More |
Specifications
Doxycycline Hydrochloride | |
10592-13-9 | |
C22H25ClN2O8 | |
Biocamycin, Tetradox | |
[Cl-].C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H]([NH+](C)C)C(=O)\C(=C(/N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12 | |
480.90 | |
480.9 |
Powder | |
Yellow | |
MFCD03427564 | |
VLUQVUWDECWBTL-UQVCFKGQSA-N | |
[(1S,4aR,11R,11aR,12S,12aR)-3-carbamoyl-4,4a,6,7,12-pentahydroxy-11-methyl-2,5-dioxo-11,11a,12,12a-tetrahydro-1H-tetracen-1-yl]-dimethylazanium;chloride | |
54706018 | |
Doxycycline hydrochloride |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Doxycycline Hydrochloride
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302 + H312-Harmful if swallowed or in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Doxycycline hydrochloride, MP Biomedicals™