Learn More
DWK Life Sciences DURAN™ Standard Cone, finished
For a rigid, self-supporting connection.
28.37 SEK - 4835.00 SEK
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
15951531
|
DWK Life Sciences
286153203 |
13 mm |
1640.00 SEK
Pack of 50 |
Estimated Shipment: 02-10-2024
Log in to see stock. |
|||||
15864578
|
DWK Life Sciences
286161606 |
56 mm |
2715.00 SEK
Pack of 10 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
15804568
|
DWK Life Sciences
286155007 |
17 mm |
2045.00 SEK
Pack of 50 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
15844558
|
DWK Life Sciences
286151005 |
6 mm |
680.00 SEK
Pack of 10 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
15854558
|
DWK Life Sciences
286152404 |
11 mm |
1835.00 SEK
Pack of 50 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
15804578
|
DWK Life Sciences
286159609 |
30 mm |
4525.00 SEK
Pack of 50 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
10181601
|
DWK Life Sciences
286160404 |
90 mm |
1250.00 SEK
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
15941531
|
DWK Life Sciences
286151802 |
8 mm |
2275.00 SEK
Pack of 50 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
15961531
|
DWK Life Sciences
286156209 |
22 mm |
2365.00 SEK
Pack of 50 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
15834578
|
DWK Life Sciences
286160207 |
40 mm |
1195.00 SEK
Pack of 10 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
15844578
|
DWK Life Sciences
286160601 |
36 mm |
1195.00 SEK
Pack of 10 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
15854578
|
DWK Life Sciences
286161203 |
46 mm |
2015.00 SEK
Pack of 10 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
15874578
|
DWK Life Sciences
286161803 |
65 mm |
4835.00 SEK
Pack of 10 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
10305211
|
DWK Life Sciences
286162002 |
80 mm |
810.00 SEK
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
15834558
|
DWK Life Sciences
286150403 |
4 mm |
690.00 SEK
Pack of 10 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
10191981
|
DWK Life Sciences
286161409 |
50 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10706941
|
DWK Life Sciences
286157008 |
26 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
The conical ground joint is the most common joint in glass apparatus engineering. Ground DURAN™ cores comply with DIN 12249 Form B in their design and meet higher requirements for surface roughness. This reduces the need for grinding grease and the risk of jamming. Ground joints with ground grease or PTFE sleeves should always be used. KECK™ Conical ground joint clamps are the perfect supplement for securing the conical ground joint.
- According to DIN 12249 Form B
- Ground (finer grinding quality, less roughness depth)
- The finer surface reduces the risk of jamming the cuts.
- Minimized grease requirement
- Tight closure due to standard ground joint
- Glass articles for further processing